Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-POMZP3 Mouse Monoclonal Antibody [clone: 2E7]
Catalog # USBI131610
Supplier:  US Biological
Anti-POMZP3 Mouse Monoclonal Antibody [clone: 2E7]
Catalog # USBI131610
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    POM121 and ZP3 fusion
  • Antigen symbol:
    POMZP3
  • Clonality:
    Monoclonal
  • Clone:
    2E7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Amino acid number:
    64 to 116
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    DQIFLDGQENKRSCLVDGLTDASSAFKVPRPGPDTLQFTVDVFHFANDSRNM*
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: POMZP3 (POM121 and ZP3 fusion)
Clonality: monoclonal
Clone: 20000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat