Order Entry
United Kingdom
ContactUsLinkComponent
Anti-MD1 Mouse Monoclonal Antibody [clone: 1H3]
Anti-MD1 Mouse Monoclonal Antibody [clone: 1H3]
Catalog # USBI129514
Supplier:  US Biological
Anti-MD1 Mouse Monoclonal Antibody [clone: 1H3]
Catalog # USBI129514
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MD1
  • Clonality:
    Monoclonal
  • Clone:
    1H3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Mouse
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    AWPTHVVCSDSGLEVLYQSCDPLQDFGFSVEKCSKQLKSNINIRFGIILREDIKELFLDLALMSQGSSVLNFSYPICEAALPKFSFCGRRKGEQIYYAGP
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: MD1
Clonality: monoclonal
Clone: 1H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse