Anti-RRP8 Mouse Polyclonal Antibody
Catalog # USBI128791
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ribosomal RNA processing 8, methyltransferase, homolog (yeast)
- Antigen symbol:RRP8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 456
- Storage buffer:PBS, pH 7,2.
- Sequence:MFEEPEWAEAAPVAAGLGPVISRPPPAASSQNKGSKRRQLLATLRALEAASLSQHPPSLCISDSEEEEEERKKKCPKKASFASASAEVGKKGKKKCQKQGPPCSDSEEEVERKKKCHKQALVGSDSAEDEKRKRKCQKHAPINSAQHLDNVDQTGPKAWKGSTTNDPPKQSPGSTSPKPPHTLSRKQWRNRQKNKRRCKNKFQPPQVPDQAPAEAPTEKTEVSPVPRTDSHEARAGALRARMAQRLDGARFRYLNEQLYSGPSSAAQRLFQEDPEAFLLYHRGFQSQVKKWPLQPVDRIARDLRQRPASLVVADFGCGDCRLASSIRNPVHCFDLASLDPRVTVCDMAQVPLEDESVDVAVFCLSLMGTNIRDFLEEANRVLKPGGLLKVAEVSSRFEDVRTFLRAVTKLGFKIVSKDLTNSHFFLFDFQKTGPPLVGPKAQLSGLQLQPCLYKRR
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RRP8 (ribosomal RNA processing 8, methyltransferase, homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human