Order Entry
United Kingdom
ContactUsLinkComponent
Anti-GLYCTK Mouse Monoclonal Antibody [clone: 1B5]
Anti-GLYCTK Mouse Monoclonal Antibody [clone: 1B5]
  USBI127386
Anti-GLYCTK Mouse Monoclonal Antibody [clone: 1B5]
  USBI127386

 

  • Antibody type:
    Primary
  • Antigen name:
    Glycerate kinase
  • Antigen symbol:
    GLYCTK
  • Clonality:
    Monoclonal
  • Clone:
    1B5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Amino acid number:
    332 to 431
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    VLSAAMQGDVKSMAQFYGLLAHVARTRLTPSMAGASVEEDAQLHELAAELQIPDLQLEEALETMAWGRGPVCLLAGGEPTVQLQGSGRGGRNQELALRV
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: GLYCTK (glycerate kinase)
Clonality: monoclonal
Clone: 1B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse