Anti-G Protein-Coupled Receptor 175 Rabbit Polyclonal Antibody
USBI127062
- Antibody type:Primary
- Antigen name:G Protein Coupled Receptor 175
- Antigen symbol:GPR175
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:Serum
- Form:liquid
- Sequence:MDTLEEVTWANGSTALPPPLAPNISVPHRCLLLLYEDIGTSRVRYWDLLLLIPNVLFLIFLLWKLPSARAKIRITSSPIFITFYILVFVVALVGIARAVVSMTVSTSNAATVADKILWEITRFFLLAIELSVIILGLAFGHLESKSSIKRVLAITTVLSLAYSVTQGTLEILYPDAHLSAEDFNIYGHGGRQFWLVSSCFFFLVYSLVVILPKTPLKERISLPSRRSFYVYAGILALLNLLQGLGSVLLCFDIIEGLCCVDATTFLYFSFFAPLIYVAFLRGFFGSEPKILFSYKCQVDETEEPDVHLPQPYAVARREGLEAAGAAGASAASYSSTQFDSAGGVAYLDDIASMPCHTGSINSTDSERWKAINA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: G Protein-Coupled Receptor 175
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human