Anti-FAP Mouse Polyclonal Antibody
Catalog # USBI126624
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast activation protein alpha
- Antigen symbol:FAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 760
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FAP (fibroblast activation protein, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human