Anti-DDT Mouse Monoclonal Antibody [clone: 1G1]
Catalog # USBI125720
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:D-dopachrome tautomerase
- Antigen symbol:DDT
- Clonality:Monoclonal
- Clone:1G1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:SandwichELISA:Thedetectionlimitis~3ng/mlasacaptureantibody
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: DDT (D-dopachrome tautomerase)
Clonality: monoclonal
Clone: 1G1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human