Anti-CCNG1 Mouse Polyclonal Antibody
Catalog # USBI125543
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cyclin G1
- Antigen symbol:CCNG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 295
- Storage buffer:PBS, pH 7,2.
- Sequence:MIEVLTTTDSQKLLHQLNALLEQESRCQPKVCGLRLIESAHDNGLRMTARLRDFEVKDLLSLTQFFGFDTETFSLAVNLLDRFLSKMKVQPKHLGCVGLSCFYLAVKSIEEERNVPLATDLIRISQYRFTVSDLMRMEKIVLEKVCWKVKATTAFQFLQLYYSLLQENLPLERRNSINFERLEAQLKACHCRIIFSKAKPSVLALSIIALEIQAQKCVELTEGIECLQKHSKINGRDLTFWQELVSKCLTEYSSNKCSKPNVQKLKWIVSGRTARQLKHSYYRITHLPTIPEMVP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: CCNG1 (cyclin G1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human