Anti-CIDEA Mouse Monoclonal Antibody [clone: 4B9]
Catalog # USBI125003
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cell death-inducing DFFA-like effector a
- Antigen symbol:CIDEA
- Clonality:Monoclonal
- Clone:4B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MRGDRASGGPGNHNGSWAREGPRLGPSWKRGLWSPRGGPNRPAEPSRPLTFMGSQTKRVLFTPLMHPARPFRVSNHDRSSRRGVMASSLQELISKTLDALVIATGLVTLVLEEDGTVVDTEEFFQTLGDNTHFMILEKGQKWMPGSQHVPTCSPPKRSGIARVTFDLYRLNPKDFIGCLNVKATMYEMYSVSYDIRCTGLKGLLRSLLRFLSYSAQVTGQFLIYLGTYMLRVLDDKEERPSLRSQAKGRFTCG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CIDEA (cell death-inducing DFFA-like effector a)
Clonality: monoclonal
Clone: 4B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human