Anti-CAST Mouse Monoclonal Antibody [clone: 2C5]
Catalog # USBI124368
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:calpastatin
- Antigen symbol:CAST
- Clonality:Monoclonal
- Clone:2C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:582 to 669
- Storage buffer:PBS, pH 7,2.
- Sequence:QPDPDENKPMEDKVKEKAKAEHRDKLGERDDTIPPEYRHLLDDNGQDKPVKPPTKKSEDSKKPADDQDPIDALSGDLDSCPSTTETSQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: CAST (calpastatin)
Clonality: monoclonal
Clone: 2C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse