Order Entry
United Kingdom
ContactUsLinkComponent
Anti-PIDD Rabbit Polyclonal Antibody
Anti-PIDD Rabbit Polyclonal Antibody
Catalog # USBI045149
Supplier:  US Biological
Anti-PIDD Rabbit Polyclonal Antibody
Catalog # USBI045149
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    p53-induced death domain Protein
  • Antigen symbol:
    PIDD
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Amino acid number:
    776 to 910
  • Storage buffer:
    PBS, 10% glycerol, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: PIDD (p53-induced death domain protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: