Anti-PLB1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
USBI040133-AP
- Antibody type:Primary
- Antigen name:phospholipase B1
- Antigen symbol:PLB1
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Thr17
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Dot blot 1:500
Do not freeze!
Type: Primary
Antigen: PLB1 (phospholipase B1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human