Anti-GIMAP8 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI036040-BIOTIN
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:GTPase, IMAP family member 8
- Antigen symbol:GIMAP8
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Amino acid number:204 to 233
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: GIMAP8 (GTPase, IMAP family member 8)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human