Anti-B4GALT5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
USBI032365-AP
- Antibody type:Primary
- Antigen name:UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 5
- Antigen symbol:B4GALT5
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Form:liquid
- Amino acid number:331 to 361
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: B4GALT5 (UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 5)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse