Anti-ATF4 Rabbit Polyclonal Antibody
Catalog # USBI032162
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Activating transcription factor 4
- Antigen symbol:ATF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Ser245
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
- Recommended dilution: ELISA1:1000 Dot blot 1:500
Type: Primary
Antigen: ATF4 (activating transcription factor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human