Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-NCOA4 Mouse Monoclonal Antibody [clone: 439CT10.4.4]
  USBI031153
undefined
Anti-NCOA4 Mouse Monoclonal Antibody [clone: 439CT10.4.4]
  USBI031153

 

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear receptor coactivator 4
  • Antigen symbol:
    NCOA4
  • Clonality:
    Monoclonal
  • Clone:
    439CT10.4.4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: NCOA4 (nuclear receptor coactivator 4)
Clonality: Monoclonal
Clone: 439CT10.4.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human