Anti-MED14 Mouse Monoclonal Antibody [clone: 483CT11.1.1]
Catalog # USBI031118
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mediator Complex Subunit 14
- Antigen symbol:MED14
- Clonality:Monoclonal
- Clone:483CT11.1.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-8000
Type: Primary
Antigen: MED14 (mediator complex subunit 14)
Clonality: Monoclonal
Clone: 483CT11.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse