Anti-UTF1 Mouse Monoclonal Antibody [clone: 13B792]
USBI030714
- Antibody type:Primary
- Antigen name:undifferentiated embryonic cell transcription factor 1
- Antigen symbol:UTF1
- Clonality:Monoclonal
- Clone:13B792
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBSwith 0,05% sodium azide and 0,5% prote
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: UTF1 (undifferentiated embryonic cell transcription factor 1)
Clonality: Monoclonal
Clone: 13B792
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human