Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-ALDH Rabbit Polyclonal Antibody
  ROCK100-4144
undefined
Anti-ALDH Rabbit Polyclonal Antibody
  ROCK100-4144

 

  • Antibody type:
    Primary
  • Antigen name:
    Aldehyde Dehydrogenase
  • Antigen symbol:
    ALDH
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Yeast
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Format:
    Antiserum
  • Form:
    Lyophilized
  • Gene ID:
    855206
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    85 mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    Aldehyde Dehydrogenase (Yeast)
  • Purification:
    delipidation and defibrination
  • Size:
    2 ml
  • Pk:
    2 mL

 

 

This product has been assayed against 1.0 µg of Aldehyde Dehydrogenase (Yeast) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ALDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast