Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-CUL7 Rabbit Monoclonal Antibody (Unconjugated) [clone: 18E1]
  ROCK100-401-A06
undefined
Anti-CUL7 Rabbit Monoclonal Antibody (Unconjugated) [clone: 18E1]
  ROCK100-401-A06

 

  • Antibody type:
    Primary
  • Antigen name:
    cullin 7
  • Antigen symbol:
    CUL7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Format:
    Antiserum
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    9820
  • Antigen synonyms:
    KIAA0076|dJ20C7.5|3M1
  • Amino acid number:
    1679 to 1698
  • Storage buffer:
    0,01% (w/v) Sodium Azide
  • Storage temperature:
    Cool
  • Concentration:
    85 mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 1679-1698 of Human Cul7 (C-terminus) coupled to KLH.
  • Purification:
    delipidation and defibrination
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

This antibody reacts with human and mouse Cul7 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and also immunoprecipitates Protein from cell lysates (using HeLa, NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CUL7
Clonality: Monoclonal
Clone: 18E1
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human