Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Calbindin D-28K (Ab-1) Rabbit Polyclonal Antibody
Anti-Calbindin D-28K (Ab-1) Rabbit Polyclonal Antibody
Catalog # PC253L-100
Anti-Calbindin D-28K (Ab-1) Rabbit Polyclonal Antibody
Catalog # PC253L-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calbindin D-28K
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Calbindin D-28K (Ab-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: