Anti-MMP-1 (Ab-1) (41-1E5) Mouse Monoclonal Antibody
IM35-100
- Antibody type:Primary
- Antigen symbol:MMP-1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: MMP-1 (Ab-1) (41-1E5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human