Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
GENSA01428-100
- Antibody type:Tag
- Antigen name:DYKDDDDK tag
- Antigen symbol:DYKDDDDK
- Clonality:Monoclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: