Anti-Serotonin receptor 1B / HTR1B Goat Polyclonal Antibody
EVBIEB07259
- Antibody type:Primary
- Antigen name:Serotonin Receptor 1B
- Antigen symbol:5-HT1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: Serotonin receptor 1B / HTR1B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: