Anti-COP1 / PSEUDO-ICE (Isofom 2) Goat Polyclonal Antibody
EVBIEB05255
- Antibody type:Primary
- Antigen name:Pseudo-ICE (Isofom 2)
- Antigen symbol:COP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: COP1 / PSEUDO-ICE (Isofom 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human