Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SHH Rabbit Polyclonal Antibody
Anti-SHH Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-SHH Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP2-22139
  • NOVUNBP2-22139SS
  • Antigen symbol:
  • SHH
  • SHH
  • Antigen name:
  • Sonic Hedgehog
  • Sonic Hedgehog
  • Antigen synonyms:
  • TPT|HLP3|HHG1|SMMCI|HPE3|MCOPCB5|TPTPS
  • TPT|HLP3|HHG1|SMMCI|HPE3|MCOPCB5|TPTPS
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 6469
  • 6469
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Frequently Bought Together