Anti-ABCG8 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 1B10A5]
NOVUNBP1-71706H
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family G, member 8
- Antigen symbol:ABCG8
- Clonality:Monoclonal
- Clone:1B10A5
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:64241
- Antigen synonyms:STSL|ATP-binding cassette sub-family G member 8|Sterolin-2|ABCG8|GBD4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: ABCG8 (ATP-binding cassette, sub-family G (WHITE), member 8)
Clonality: monoclonal
Clone: 1B10A5
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse