Anti-GATA4 Mouse monoclonal antibody unconjugated [clone: 6H10]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: GATA4 (GATA binding protein 4)
Clonality: monoclonal
Clone: 6H10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- NOVUNBP1-47540
- NOVUNBP1-47540SS
- Antigen symbol:
- GATA4
- GATA4
- Antigen name:
- GATA binding Protein 4
- GATA binding protein 4
- Antigen synonyms:
- GATA-4|Transcription factor GATA-4|DNA binding protein GATA-GT2|GAT4|GATA binding factor-4
- ASD2, TACHD, TOF, VSD1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 6H10
- 6H10
- Reactivity:
- Human
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 2626
- 2626
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together