Anti-Adropin Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNBP1-26387C
- Antibody type:Primary
- Antigen name:Adropin
- Antigen symbol:Adropin
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:375704
- Antigen synonyms:C9orf165|UNQ470
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: Adropin (adropin)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse