Anti-Luciferase Mouse Monoclonal Antibody [clone: Luci 21 1-107]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: Luciferase
Clonality: monoclonal
Clone: Luci 21 1-107
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Specifications
- Catalog No:
- NOVUNB600-307SS
- NOVUNB600-307
- Antigen name:
- Luciferase
- Luciferase
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Luci 21 1-107
- Luci 21 1-107
- Reactivity:
- Firefly
- Firefly
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together