Anti-APOE Mouse monoclonal antibody unconjugated [clone: WUE-4]
Supplier: Novus Biologicals
Certificates
About this item
The Apolipoprotein E / ApoE Antibody (WUE-4) from Novus Biologicals is a mouse monoclonal antibody to Apolipoprotein E / ApoE. This antibody reacts with human, mouse, rat (negative). The Apolipoprotein E / ApoE Antibody (WUE-4) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
CyTOF-ready
Type: Primary
Antigen: APOE
Clonality: Monoclonal
Clone: WUE-4
Conjugation: Unconjugated
Epitope: Residues 140-160 of purified human ApoE (LDLR receptor binding site).
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat (Negative)
Specifications
- Catalog No:
- NOVUNB110-60531
- NOVUNB110-60531SS
- Antigen symbol:
- APOE
- APOE
- Antigen name:
- Apolipoprotein E
- Apolipoprotein E
- Antigen synonyms:
- apo-E|AD2|LDLCQ5|LPG|MGC1571
- apo-E|AD2|LDLCQ5|LPG|MGC1571
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- WUE-4
- WUE-4
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 348
- 348
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together