Anti-WT1 Mouse Monoclonal Antibody (Biotin) [clone: 6F-H2]
Catalog # NOVUNB110-60011B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Wilms tumor 1
- Antigen symbol:WT1
- Clonality:Monoclonal
- Clone:6F-H2
- Conjugation:Biotin
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:7490
- Antigen synonyms:WAGR|WT33|WIT-2|WT1|GUD|Wilms tumor 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: WT1 (Wilms tumor 1)
Clonality: monoclonal
Clone: 6F-H2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human