Anti-TMED10 Rabbit Polyclonal Antibody (Biotin)
NOVUNB110-57586B
- Antibody type:Primary
- Antigen name:Transmembrane emp24 domain-containing protein 10
- Antigen symbol:TMED10
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:10972
- Antigen synonyms:TMED10|Tmp 21 I|21 kDa transmembrane trafficking protein|Transmembrane protein Tmp21|TMED 10|Transmembrane emp24 domain containing protein 10|S31III125|p23|Transmembrane trafficking protein 21kD|S31I125|1110014C03Rik|Transmembrane emp24-like trafficking protein 10 yeast|MGC102351|p24delta|p24delta1|TMEDA_HUMAN.|p24 family protein delta 1|Tmp 21
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: TMED10 (transmembrane emp24-like trafficking protein 10 (yeast))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: