Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CITED4 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: HT13-2D6.3]
Anti-CITED4 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: HT13-2D6.3]
  NOVUNB110-41572F
Anti-CITED4 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: HT13-2D6.3]
  NOVUNB110-41572F

 

  • Antibody type:
    Primary
  • Antigen name:
    Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4
  • Antigen symbol:
    CITED4
  • Clonality:
    Monoclonal
  • Clone:
    HT13-2D6.3
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    163732
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: CITED4 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4)
Clonality: monoclonal
Clone: HT13-2D6.3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human