Anti-BMI1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
NOVUNB110-40823F
- Antibody type:Primary
- Antigen name:BMI1 proto-oncogene, polycomb ring finger
- Antigen symbol:BMI1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:648
- Antigen synonyms:flvi-2/bmi-1|RNF51|FLVI2/BMI1|PCGF4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: BMI1 (BMI1 polycomb ring finger oncogene)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse