Anti-CA9 Rabbit Polyclonal Antibody
Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
- Catalog No:
- NOVUNB100-417SS
- NOVUNB100-417
- Antigen symbol:
- CA9
- CA9
- Antigen name:
- Carbonic Anhydrase 9
- Carbonic Anhydrase 9
- Antigen synonyms:
- CAIX|carbonic dehydratase|membrane antigen MN|carbonic anhydrase 9|RCC-associated protein G250|MN|carbonate dehydratase IX|CA-IX|RCC-associated antigen G250|P54/58N|pMW1|renal cell carcinoma-associated antigen G250
- CAIX|carbonic dehydratase|membrane antigen MN|carbonic anhydrase 9|RCC-associated protein G250|MN|carbonate dehydratase IX|CA-IX|RCC-associated antigen G250|P54/58N|pMW1|renal cell carcinoma-associated antigen G250
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 768
- 768
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Gel shift:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together