Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-RAD21 Mouse Monoclonal Antibody (DyLight® 550) [clone: CM110-2C10]
  NOVUNB100-386R
Anti-RAD21 Mouse Monoclonal Antibody (DyLight® 550) [clone: CM110-2C10]
  NOVUNB100-386R

 

  • Antibody type:
    Primary
  • Antigen name:
    RAD21 homolog
  • Antigen symbol:
    RAD21
  • Clonality:
    Monoclonal
  • Clone:
    CM110-2C10
  • Conjugation:
    DyLight® 550
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    5885
  • Antigen synonyms:
    SCC1|hHR21|KIAA0078|FLJ40596|RAD21|MCD1|NXP1|FLJ25655|HR21|NXP-1|HRAD21
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: RAD21 (RAD21 homolog (S. pombe))
Clonality: monoclonal
Clone: CM110-2C10
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse