Anti-CASR Mouse Monoclonal Antibody (DyLight® 650) [clone: HL 1499]
NOVUNB100-1830C
- Antibody type:Primary
- Antigen name:Calcium Sensing Receptor
- Antigen symbol:CASR
- Clonality:Monoclonal
- Clone:HL 1499
- Conjugation:DyLight® 650
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:846
- Antigen synonyms:Parathyroid Cell calcium-sensing receptor|EIG8|FHH|GPRC2A|Extracellular calcium-sensing receptor precursor|FIH|HYPOC1|HHC1|PCAR1|CAR|HHC|NSHPT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: CASR (calcium-sensing receptor)
Clonality: monoclonal
Clone: HL 1499
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: