Anti-liver Arginase Rabbit Polyclonal Antibody
Catalog # BSBTPA1783
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:liver Arginase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:307 - 323
- Sequence:KREGNHKPETDYLKPPK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of rat liver Arginase (307-323aa KREGNHKPETDYLKPPK), different from the related mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Arginase-1 (ARG1) detection. Tested with WB in Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: liver Arginase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat