Anti-CDK4 Mouse Monoclonal Antibody [clone: IML-4]
Catalog # BSBTMA1020
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent Kinase 4
- Antigen symbol:CDK4
- Clonality:Monoclonal
- Clone:IML-4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:CMM3|MGC14458|PSK-J3|Melanoma Cutaneous Malignant 3|Cell Division Kinase 4|Cell Division Protein Kinase 4
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant human Cdk4 protein.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CDK4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat