Anti-Cytochrome C Oxidase subunit VIb Mouse Monoclonal Antibody [clone: 3F9D3D11AF6]
Catalog # ABCAAB110266-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome C Oxidase subunit VIb
- Clonality:Monoclonal
- Clone:3F9D3D11AF6
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Cytochrome C Oxidase subunit VIb
Clonality: monoclonal
Clone: 3F9D3D11AF6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: