Order Entry
United Kingdom
ContactUsLinkComponent
Anti-NETO2 Rabbit Monoclonal Antibody [clone: EPR3497]
Anti-NETO2 Rabbit Monoclonal Antibody [clone: EPR3497]
Catalog # ABCAAB109288-100
Supplier:  Abcam
Anti-NETO2 Rabbit Monoclonal Antibody [clone: EPR3497]
Catalog # ABCAAB109288-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neuropilin (NRP) and tolloid (TLL)-like 2
  • Antigen symbol:
    NETO2
  • Clonality:
    Monoclonal
  • Clone:
    EPR3497
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: NETO2 (neuropilin (NRP) and tolloid (TLL)-like 2)
Clonality: monoclonal
Clone: EPR3497
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: