Anti-APOA4 Mouse Monoclonal Antibody
Catalog # BIRBORB89242-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein A4
- Antigen symbol:APOA4
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites fluid
- Storage buffer:0,03% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ELISA: 1:10000; Immunochemistry: 1:200-1:400; Western Blot: 1:500-1:2000
Type: Primary
Antigen: APOA4 (apolipoprotein A-IV)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human