Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-C. trachomatis EB Rabbit Polyclonal Antibody (Biotin)
  BIRBORB23149-1
 :  Biorbyt
Anti-C. trachomatis EB Rabbit Polyclonal Antibody (Biotin)
  BIRBORB23149-1
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    C. trachomatis EB
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Storage buffer:
    0,01M PBS, pH 7,2No stabilizing proteins have been added.
  • Storage temperature:
    Cool
  • Concentration:
    4-5 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    L2 + other serovar groups
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Primary
Antigen: C. trachomatis EB
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: