Anti-IgG Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21658-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 2,7, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified Fab of normal IgG isolated form rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: None
Isotype: none
Reactivity: