Anti-PTF1A Goat Polyclonal Antibody
BIRBORB19531-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Pancreas Specific Transcription Factor 1a
- Antigen symbol:PTF1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Molecular weight:48 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:internal region
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
ELISA: 1:32000; Western Blot: 0.5-1.5 ?g/ml
Type: Primary
Antigen: PTF1A (pancreas specific transcription factor, 1a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: