Anti-SP1 Goat Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
ELISA: 1:16000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: SP1 (Sp1 transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Specifications
- Catalog No:
- BIRBORB19050-100
- BIRBORB19050-20
- Antigen symbol:
- SP1
- SP1
- Antigen name:
- Sp1 transcription factor
- Sp1 transcription factor
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Goat
- Goat
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- internal region
- internal region
- Molecular weight:
- 100 kDa
- 100 kDa
- Purification:
- purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Storage buffer:
- Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together