Anti-APITD1 Mouse Polyclonal Antibody
ABNOH00378708-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Apoptosis-Inducing, TAF9-Like Domain 1
- Antigen symbol:APITD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:378708
- Antigen synonyms:CENPS|CENP-S|FAAP16|MHF1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: APITD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human