Anti-FRMD3 Mouse Monoclonal Antibody [clone: 2F1]
Catalog # ABNOH00257019-M09
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:FERM domain containing 3
- Antigen symbol:FRMD3
- Clonality:Monoclonal
- Clone:2F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:257019
- Antigen synonyms:EPB41L4O|EPB41LO|4.1O|P410
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FRMD3
Clonality: Monoclonal
Clone: 2F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human