Anti-TP53RK Mouse Monoclonal Antibody [clone: 2A9]
Catalog # ABNOH00112858-M02A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TP53 regulating kinase
- Antigen symbol:TP53RK
- Clonality:Monoclonal
- Clone:2A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:112858
- Antigen synonyms:PRPK|C20orf64|BUD32|dJ101A2|Nori-2|Nori-2p
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: TP53RK
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human